5-Bromo-4-methyl-2-phenyl-pyridine ≥95%

Supplier: AOBChem USA

Warning

25716-5G 25716-1G 25716-25G 25716-500MG
77632-284EA 841 USD
77632-284 77637-504 77654-470 77649-558
5-Bromo-4-methyl-2-phenyl-pyridine ≥95%
5-Bromo-4-methyl-2-phenyl-pyridine

Caution: For Research Use Only

Formula: C₁₂H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD23713141
CAS Number: 31686-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR