(5-Bromo-2-methoxyphenyl)-N,N-dimethylmethanamine ≥97%

Supplier: AOBChem USA

Warning

24494-5G 24494-1G 24494-25G 24494-500MG
77634-466EA 425 USD
77634-466 77637-418 77658-178 77649-460
(5-Bromo-2-methoxyphenyl)-N,N-dimethylmethanamine ≥97%
(5-Bromo-2-methoxyphenyl)-N,N-dimethylmethanamine

Caution: For Research Use Only

Formula: 244.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02640922
CAS Number: 7078-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR