α-Ethenyl-2-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234941-5G 234941-1G 234941-500MG
77628-172EA 1609.89 USD
77628-172 77645-354 77645-356
α-Ethenyl-2-ethylbenzenemethanol ≥95%
α-Ethenyl-2-ethylbenzenemethanol

Caution: For Research Use Only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21232159
CAS Number: 1071201-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR