4-(4-Methylthiophenyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

22900-5G 22900-25G 22900-1G 22900-500MG
77634-422EA 326 USD
77634-422 77658-084 77641-304 77649-342
4-(4-Methylthiophenyl)phenylboronic acid ≥95%
4-(4-Methylthiophenyl)phenylboronic acid

Caution: For Research Use Only

Formula: C₁₃H₁₃BO₂S
Storage Temperature: Ambient
MDL Number: MFCD08701730
CAS Number: 501944-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR