5-Chloro-2-fluoro-N-methoxy-N-methyl-3-pyridinecarboxamide ≥97%

Supplier: AOBChem USA

Warning

21054-5G 21054-25G 21054-1G 21054-500MG
77632-000EA 841 USD
77632-000 77657-976 77641-204 77645-198
5-Chloro-2-fluoro-N-methoxy-N-methyl-3-pyridinecarboxamide ≥97%
5-Chloro-2-fluoro-N-methoxy-N-methyl-3-pyridinecarboxamide

Caution: For Research Use Only

Formula: C₈H₈ClFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18257899
CAS Number: 1256834-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR