2'-Bromo-6'-fluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Bromo-6-fluorophenyl)ethanone

18453-5G 18453-25G
77634-234EA 87 USD
77634-234 77657-790
2'-Bromo-6'-fluoroacetophenone ≥95%
2'-Bromo-6'-fluoroacetophenone

Caution: For Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD13193629
CAS Number: 928715-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR