2-Chloro-3-fluoro-1,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

179734-5G 179734-25G 179734-1G 179734-500MG
77627-954EA 1484.23 USD
77627-954 77653-938 77644-988 77649-018
2-Chloro-3-fluoro-1,4-dimethylbenzene ≥95%
2-Chloro-3-fluoro-1,4-dimethylbenzene

Caution: For Research Use Only

Formula: C₈H₈ClF
Storage Temperature: Ambient
MDL Number: MFCD28751488
CAS Number: 1805224-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR