4-Bromo-3-(trifluoromethoxy)benzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
16978-5G
16978-1G
16978-25G
16978-500MG
77631-794EA
625
USD
77631-794
77636-924
77653-838
77648-934
4-Bromo-3-(trifluoromethoxy)benzoic acid ≥95%
4-Bromo-3-(trifluoromethoxy)benzoic acid
Caution: For Research Use Only
Formula:
C₈H₄BrF₃O₃ Storage Temperature: Ambient |
MDL Number:
MFCD22378818 CAS Number: 1008769-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...