3-(3-Chloro-2-fluorophenyl)-3-oxetanol ≥97%
Supplier: AOBChem USA
|
Warning
|
166251-1G
166251-5G
166251-500MG
77636-806EA
507
USD
77636-806
77653-742
77644-782
3-(3-Chloro-2-fluorophenyl)-3-oxetanol ≥97%
3-(3-Chloro-2-fluorophenyl)-3-oxetanol
Caution: For Research Use Only
Formula:
C₉H₈ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD34851804 CAS Number: 2583023-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...