(5-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
165854-5G
165854-1G
165854-500MG
165854-25G
77627-838EA
1553.24
USD
77627-838
77636-796
77652-384
77657-592
(5-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane
Caution: For Research Use Only
Formula:
C₉H₁₀BrFS Storage Temperature: Ambient |
MDL Number:
MFCD34851537 CAS Number: 2921864-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...