2,2-Difluoro-1-(4-(tert-pentyl)phenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
158004-5G
158004-25G
158004-500MG
158004-1G
77631-374EA
1797.35
USD
77631-374
77657-524
77652-364
77644-704
2,2-Difluoro-1-(4-(tert-pentyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(4-(tert-pentyl)phenyl)ethanone
Caution: For Research Use Only
Formula:
C₁₃H₁₆F₂O Storage Temperature: Ambient |
MDL Number:
MFCD34846482 CAS Number: 1697892-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...