(3-(Cyclopropylmethoxy)-2,4-difluorophenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
148558-5G
148558-25G
148558-1G
148558-500MG
77631-660EA
1441
USD
77631-660
77657-482
77640-686
77648-708
(3-(Cyclopropylmethoxy)-2,4-difluorophenyl)(methyl)sulfane ≥95%
(3-(Cyclopropylmethoxy)-2,4-difluorophenyl)(methyl)sulfane
Caution: For Research Use Only
Formula:
C₁₁H₁₂F₂OS Storage Temperature: Ambient |
MDL Number:
MFCD34840158 CAS Number: 2921902-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...