1-(2,3-Dichloropyridin-4-yl)-2-methylpropan-1-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
118259-5G
118259-25G
118259-1G
118259-500MG
77631-104EA
1484.23
USD
77631-104
77657-242
77640-424
77644-408
1-(2,3-Dichloropyridin-4-yl)-2-methylpropan-1-ol ≥97%
1-(2,3-Dichloropyridin-4-yl)-2-methylpropan-1-ol
Caution: For Research Use Only
Formula:
C₉H₁₁Cl₂NO Storage Temperature: Ambient |
MDL Number:
MFCD34784008 CAS Number: 2918863-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...