AZD0780 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6'-(((1S,3S)-3-((5-(Difluoromethoxy)pyrimidin-2-yl)amino)cyclopentyl)amino)-2H-[1,3'-bipyridin]-2-one
A1937165-1MG
A1937165-100MG
A1937165-10MG
A1937165-25MG
A1937165-5MG
77625-942EA
97.85
USD
77625-942
77877-482
77877-483
77877-484
77877-485
AZD0780 98%
AZD0780
Caution: for Research Use Only
Formula:
C₂₀H₂₀F₂N₆O₂ MW: 414.41 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD34676592 CAS Number: 2455427-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...