Nefiracetam ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: N-(2,6-Dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide

N0996-1G N0996-200MG
TCN0996-1GEA 598 USD
TCN0996-1G TCN0996-200MG
Nefiracetam ≥98.0% (by total nitrogen basis)
Nefiracetam
Formula: C₁₄H₁₈N₂O₂
MW: 246.31 g/mol
MDL Number: MFCD00209882
CAS Number: 77191-36-7
Merck Index: 14,06439

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR