(4-Nitrophenyl)urea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

N0980-1G N0980-5G
TCN0980-1GEA 96.2 USD
TCN0980-1G TCN0980-5G
(4-Nitrophenyl)urea ≥98.0% (by HPLC, total nitrogen)
(4-Nitrophenyl)urea
Formula: C₇H₇N₃O₃
MW: 181.15 g/mol
Melting Pt: 217°C
MDL Number: MFCD09040599
CAS Number: 556-10-5
Merck Index: 14,06626

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR