(S)-2,4-Diphenyl-4,5-dihydrooxazole ≥98.0% (by GC)

Supplier: TCI America

D5368-1G D5368-5G
76219-954EA 138 USD
76219-954 76219-956
(S)-2,4-Diphenyl-4,5-dihydrooxazole ≥98.0% (by GC)
(S)-2,4-Diphenyl-4,5-dihydrooxazole
Formula: C₁₅H₁₃NO
CAS Number: 475571-19-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR