Alimemazine Tartrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate, Alimemazine Hemitartrate

A2813-100MG A2813-1G A2813-5G
TCA2813-100MGEA 32 USD
TCA2813-100MG TCA2813-1G TCA2813-5G
Alimemazine Tartrate ≥98.0% (by HPLC, titration analysis)
Alimemazine Tartrate
Formula: C₃₆H₄₄N₄S₂·C₄H₆O₆
MW: 746.98 g/mol
MDL Number: MFCD00242594
CAS Number: 4330-99-8
EINECS: 224-368-9
UN: 2811
ADR: 6.1 III
Merck Index: 14,09704

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR