9,9-Bis(3,4-dihydroxyphenyl)fluorene ≥98.0% (by GC)

Supplier: TCI America

B4665-1G B4665-5G
TCB4665-1GEA 138 USD
TCB4665-1G TCB4665-5G
9,9-Bis(3,4-dihydroxyphenyl)fluorene ≥98.0% (by GC)
9,9-Bis(3,4-dihydroxyphenyl)fluorene
Formula: C₂₅H₁₈O₄
CAS Number: 351521-78-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 181.0 to 185.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR