2-n-Octylbenzotriazole ≥98.0% (by GC)

Supplier: TCI America

O0430-1G O0430-200MG
TCO0430-1GEA 366 USD
TCO0430-1G TCO0430-200MG
2-n-Octylbenzotriazole ≥98.0% (by GC)
2-n-Octylbenzotriazole
Formula: C₁₄H₂₁N₃
Boiling Pt: 110°C
MDL Number: MFCD00963848
CAS Number: 112642-69-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR