Beclomethasone dipropionate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Beclometasone dipropionate

B4464-1G B4464-5G
TCB4464-1GEA 144 USD
TCB4464-1G 76219-392
Beclomethasone dipropionate ≥98.0% (by HPLC)
Beclomethasone dipropionate
Formula: C₂₈H₃₇ClO₇
MDL Number: MFCD00135613
CAS Number: 5534-09-8
EINECS: 226-886-0
Merck Index: 14,01019

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +88.0 to +94.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR