4-Chloropicolinamide ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Chloropyridine-2-carboxamide
C2949-1G
C2949-5G
TCC2949-1GEA
19
USD
TCC2949-1G
TCC2949-5G
4-Chloropicolinamide ≥97.0% (by HPLC, titration analysis)
4-Chloropicolinamide
Formula:
C₆H₅ClN₂O MW: 156.57 g/mol |
MDL Number:
MFCD01085339 CAS Number: 99586-65-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...