4-Amino-o-anisic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Amino-2-methoxybenzenecarboxylic acid
A2840-25G
A2840-5G
TCA2840-25GEA
203
USD
TCA2840-25G
TCA2840-5G
4-Amino-o-anisic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-o-anisic acid
Formula:
C₈H₉NO₃ MW: 167.16 |
MDL Number:
MFCD00114479 CAS Number: 2486-80-8 EINECS: 219-632-5 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...