2-Cyclobutyl-10-methyl-3-phenylpyrimido[4,5-b]quinoline-4,5(3H,10H)-dione 98%

Supplier: Ambeed
Warning

A1550788-1MG A1550788-5MG A1550788-10MG A1550788-25MG
77598-054EA 38.95 USD
77598-054 77625-404 77876-287 77876-288
2-Cyclobutyl-10-methyl-3-phenylpyrimido[4,5-b]quinoline-4,5(3H,10H)-dione 98%
2-Cyclobutyl-10-methyl-3-phenylpyrimido[4,5-b]quinoline-4,5(3H,10H)-dione

Caution: for Research Use Only

Formula: C₂₂H₁₉N₃O₂
CAS Number: 1613509-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR