Sterile Blood Bank Pipettes
Supplier: Copan Diagnostics
212CS05
30002-060CS 214.95 USD
30002-060
Sterile Blood Bank Pipettes
Pipettes Pasteur/Transfer Pipettes
These 2 ml, graduated, Pasteur-style sterile pipettes are ideal for transferring and dispensing liquids safely in all types of laboratories, eliminating the risk of cross-contamination.
They are made of nontoxic, low-density polyethylene and can be incinerated. They can also be heat-sealed and frozen at very low temperatures such as in liquid nitrogen.
Ordering information: Sterile pipettes are packaged in medical-grade paper and atoxic film. Packed five pipettes per pack; case contains 100 packs.