Fendall® Saline Eyewash Wall Stations, Honeywell Safety
Supplier: Honeywell Safety Products
32-000461-0000 32-000460-0000-H5
56611-128CS 1063.22 USD
56611-128 56611-130 56611-126
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
First Aid Supplies Eyewash Stations and Eyewash Bottles
Available in 470 ml (16 oz.) and 950 ml (32 oz.) single and 950 ml (32 oz.) double wall stations.
  • Two-sided tape and screws are provided for mounting
  • Classified as personal/secondary eyewash device
Bottles have extended flow openings and twist-off tops for easy access in an emergency. Each bottle is expiry dated and lot numbered for end-user protection.