Temperature Calibration Bowls
Supplier: Sartorius
YTM09MA YTM10MA
76413-066EA 905.8 USD
76413-066 76413-068
Temperature Calibration Bowls
Balances and Scales
Designed for use with Mark 3 Series moisture analyzers. Provides automatic calibartions.
The temperature calibration bowl provides automatic calibrations for your moisture analyzer. To calibrate the internal temperature sensor, simply remove the tray support and breeze shield wihin the heating chamber, and install the bowl.
Delivery information: With one year warranty.