Hydrometer Racks
Supplier: Heathrow Scientific
121002 121003
77591-822CS 520.51 USD
77591-822 77591-824
Hydrometer Racks
Racks Hydrometer Racks
These hydrometer racks accomdate up to 10 hydrometers with diameters up to 31 mm, ensuring safe storage, efficient drying, and secure transport.
  • Teardrop-shaped slots hold hydrometers securely in place, support a range of diameters, and ensure easy insertion/removal for convenient handling
  • 3-Tier setup provides enhanced stability and protection of hydrometer body
  • Built-in handles provide a secure grip for safe and convenient transport
  • Removable drip tray simplifies cleaning and maintenance, improving sanitation and protecting your workspace
  • Stable wide
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable