HOBO® Solar Radiation Shield
Supplier: ONSET COMPUTER CORPORATION SE
RS3-B
470338-048EA 102.53 USD
470338-048
HOBO® Solar Radiation Shield
Shields Radiation Shields
Use the solar radiation shield with HOBO external sensors for improved temperature measurement accuracy in locations exposed to sunlight.
  • Research grade data collection
  • Great for budding environmental scientists
The small size and unobstructed airflow of this shield provides a faster response to changing conditions than larger radiation shields.