IPTG (isopropyl-β-D-thiogalactopyranoside) solution 100 mM, Novagen®

Supplier: MilliporeSigma

Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triolIPTGIsopropyl Beta-D-1-thiogalactopyranoside

70527-3
EM70527-3EA 242.66 USD
EM70527-3
IPTG (isopropyl-β-D-thiogalactopyranoside) solution 100 mM, Novagen®
IPTG (Isopropyl-β-D-thiogalactopyranoside)

Convenient, ready-to-use solution

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR