L(+)-Leucine ≥98.5% (by assay), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: L-Leucine(S)-(+)-LeucineLeucineLeuH-Leu-OH

4330-100GM
80058-318EA 134.67 USD
80058-318
L(+)-Leucine ≥98.5% (by assay), Calbiochem®, Millipore®
L(+)-Leucine

L-Leucine, an essential branched chain amino acid.

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR