2-Guanidinoethylmercaptosuccinic acid ≥99% (by TLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: GEMSA

369334-5MG
80056-610EA 248.99 USD
80056-610
2-Guanidinoethylmercaptosuccinic acid ≥99% (by TLC), Calbiochem®, Sigma-Aldrich®
2-Guanidinoethylmercaptosuccinic acid

Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Kᵢ = 8.8 nM).

Formula: C₇H₁₃N₃O₄S
MW: 235.2 g/mol
Storage Temperature: Refrigerator
CAS Number: 77482-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR