EDTA trisodium salt 0.5 M, Molecular Biology Grade, DEPC treated pH 8.0, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Danger

324506-100ML
82604-002EA 103.17 USD
82604-002
EDTA trisodium salt 0.5 M, Molecular Biology Grade, DEPC treated pH 8.0, Calbiochem®, Millipore®
EDTA trisodium salt
Formula: C₁₀H₁₃N₂Na₃O₈
MW: 358.19 g/mol
Storage Temperature: Ambient
CAS Number: 150-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR