Congo red ≥75% (dry basis), high purity, Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28

234610-500MG
80511-204EA 116.62 USD
80511-204
Congo red ≥75% (dry basis), high purity, Calbiochem®, Sigma-Aldrich®
Congo Red

An amyloidophylic dye that specifically stains stacked β sheet aggregates.

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
Merck Index: 12,02562

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR