Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥95% (by HPLC) low metals, Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Adenosine-5'-triphosphoric acid disodium salt trihydrateATP disodium trihydrateATP, 2Na 3H2O(((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate5'-ATP·2Na·3H2O

119125-1GM
80050-140EA 148.83 USD
80050-140
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥95% (by HPLC) low metals, Calbiochem®, Millipore®
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) trihydrate

Low metals grade. Specially processed to reduce levels of contaminating metals. Suitable for enzyme kinetic studies.

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MW: 605.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00150754
CAS Number: 987-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR