PS Tubes with Cap, Round Base, Sterile
Supplier: SARSTEDT INC
55.468.001 55.475.001 55.484.001
77587-534CS 170.37 USD
77587-534 77587-544 77587-564
PS Tubes with Cap, Round Base, Sterile
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, it offers the right product for every laboratory and analytical requirement.
  • Material: PS
  • Graduations: No
  • Purity: Sterile
  • Closure: Assembled
  • Base shape: Round base
  • Corresponds to the requirement: Sealed at 0.6 bar/10 min
  • Polystyrene (PS) tubes for maximum transparency
  • Large grip for optimal handling
  • Excellent sealing properties
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.