Anti-CD14 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Sino Biological
90174MM12F100 90174MM12F25
77586-796EA 72.1 USD
77586-796 77586-798
Anti-CD14 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus CD14 ( Catalog#90174-C08H; B3Y6B8; Met1-Met344) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.