Anti-FCGR3 Mouse Antibody
Supplier: Sino Biological
90013-MM01-20 90013-MM05-20
77586-740EA 82.4 USD
77586-740 77586-742
Anti-FCGR3 Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus CD16 / Fc gamma RIII ( Catalog#90013-C08H; NP_001258584.1; Met1-Gly206). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.