Anti-IL17A Rabbit Antibody (PE (Phycoerythrin))
Supplier: Sino Biological
51065R217P100 51065R217P25
77586-302EA 92.7 USD
77586-302 77586-304
Anti-IL17A Rabbit Antibody (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17 (rM IL17; Catalog#51065-MNAE; Q62386; Thr22-Ala158) and conjugated with PE under optimum conditions, the unreacted PE was removed.