Anti-CD83 Rabbit Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Sino Biological
50694R101F100 50694R101F25
77586-072EA 154.5 USD
77586-072 77586-074
Anti-CD83 Rabbit Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD83 / HB15 (rM CD83 / HB15; Catalog#50694-M08H; O88324; Met1-Arg133) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.