Anti-MPXV-B6R Rabbit Antibody (HRP (Horseradish Peroxidase))
Supplier: Sino Biological
40902R006H100 40902R006H20
77585-604EA 463.5 USD
77585-604 77585-606
Anti-MPXV-B6R Rabbit Antibody (HRP (Horseradish Peroxidase))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus (MPXV) B6R (Catalog# 40902-V08H; QNP13760.1; Tyr18-His279) and conjugated with horseradish-peroxidase (HRP).