Anti-LILRB4 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Sino Biological
16742MM05F25 16742MM05F100
77584-964EA 92.7 USD
77584-964 77584-962
Anti-LILRB4 Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB4/CD85k/ILT3 (rh LILRB4/CD85k/ILT3; Catalog#16742-H08H; NP_001265357.2; Met1-Glu259) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.