Anti-TPO Mouse Antibody
Supplier: Sino Biological
13194-MM12-100 13194-MM12-20
77584-710EA 277.07 USD
77584-710 77584-712
Anti-TPO Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Thyroid peroxidase (rh Thyroid peroxidase; Catalog#13194-H08B; P07202-1; Met1-Arg846,257Ser/Ala,725Pro/Thr). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.