Anti-ACNB Mouse Antibody
Supplier: Sino Biological
13102-MM05-20 13102-MM09-20
77584-678EA 103 USD
77584-678 77584-680
Anti-ACNB Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Escherichia coli acnB ( Catalog#13102-ENAE; P36683; Met1-Val865). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.