Anti-ACNB Mouse Antibody (HRP (Horseradish Peroxidase))
Supplier: Sino Biological
13102MM02H100 13102MM02H20
77584-674EA 412 USD
77584-674 77584-676
Anti-ACNB Mouse Antibody (HRP (Horseradish Peroxidase))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Escherichia coli acnB ( Catalog#13102-ENAE; P36683; Met1-Val865). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).