Anti-CD8A Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Sino Biological
10980MM48F100 10980MM48F25
77583-260EA 30.9 USD
77583-260 77583-262
Anti-CD8A Mouse Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD8 alpha/CD8A (rh CD8 alpha/CD8A; Catalog#10980-H08H; NP_001759.3; Met1-Asp182) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.