Anti-CFH Mouse Antibody
Supplier: Sino Biological
10714-MM02-20 10714-MM03-20
77582-928EA 61.8 USD
77582-928 77582-930
Anti-CFH Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Factor H / CFH (rh Complement Factor H / CFH; Catalog#10714-H08H; NP_000177.2; Ser 860-Arg 1231). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.