Anti-BMP4 Mouse Antibody
Supplier: Sino Biological
10609-MM21-100 10609-MM27-100
77582-796EA 277.07 USD
77582-796 77582-798
Anti-BMP4 Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BMP4 (rh BMP4; Catalog#10609-HNAE2; P12644; Ser293-Arg408). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.