Anti-PDCD1 Mouse Antibody (APC (Allophycocyanin))
Supplier: Sino Biological
10377MM18A100 10377MM18A25
77582-362EA 288.4 USD
77582-362 77582-364
Anti-PDCD1 Mouse Antibody (APC (Allophycocyanin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD1 / PDCD1 (rh PD1 / PDCD1; Catalog#10377-H08H; NP_005009.2; Met1-Gln167) and conjugated with APC under optimum conditions, the unreacted APC was removed.