Anti-GAPDH Mouse Antibody
Supplier: Sino Biological
10094-MM09-100 10094-MM13-100
77581-712EA 225.57 USD
77581-712 77581-714
Anti-GAPDH Mouse Antibody
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GAPDH (rh GAPDH; Catalog#10094-H07E; P04406; Met1-Glu335). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.